Assay Design



Gene Transcripts

Gene Symbol
Mog
Entrez Gene ID
24558
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
-
Chromosome Location
Chr.20: 2003575 - 2014284 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022668.2 NP_073159.2 2 - 3 521 55
XM_006255867.2 XP_006255929.1 2 - 3 994 55
XM_008772687.1 XP_008770909.1 2 - 3 998 55
GenBank mRNA BC087717.1 - 2 - 3 521 55
FQ214199.1 - 2 - 3 501 55
L21995.1 - 1 - 2 352 55
M99485.1 - 2 - 3 521 55

View Full Product Details