Assay Design



Gene Transcripts

Gene Symbol
Amhr2
Entrez Gene ID
29530
Gene Name
anti-Mullerian hormone receptor, type II
Gene Aliases
-
Chromosome Location
Chr.7: 144052202 - 144060678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030998.1 NP_112260.1 10 - 11 1491 81
GenBank mRNA U42427.1 - 10 - 11 1491 81
X71916.1 - 10 - 11 1473 81

View Full Product Details