Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031026.1 | NP_112288.1 | 3 - 4 | 305 | 117 |
XM_006255053.2 | XP_006255115.1 | 3 - 4 | 519 | 117 | |
XM_006255054.2 | XP_006255116.1 | 3 - 4 | 519 | 117 | |
GenBank mRNA | BC089794.1 | - | 3 - 4 | 326 | 117 |
U15138.1 | - | 3 - 4 | 305 | 117 |