Assay Design



Gene Transcripts

Gene Symbol
Il1b
Entrez Gene ID
24494
Gene Name
interleukin 1 beta
Gene Aliases
-
Chromosome Location
Chr.3: 121876256 - 121882637 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031512.2 NP_113700.2 4 - 5 351 66
GenBank mRNA BC091141.1 - 4 - 5 351 66
M98820.1 - 4 - 5 371 66

View Full Product Details