Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031534.2 | NP_113722.2 | 2 - 3 | 582 | 85 |
XM_006234620.1 | XP_006234682.1 | 2 - 3 | 975 | 85 | |
XM_006234621.2 | XP_006234683.1 | 2 - 3 | 1053 | 85 | |
XM_006234622.2 | XP_006234684.1 | 2 - 3 | 1053 | 85 | |
GenBank mRNA | X69716.1 | - | 2 - 3 | 582 | 85 |