Assay Design



Gene Transcripts

Gene Symbol
Add3
Entrez Gene ID
25230
Gene Name
adducin 3 (gamma)
Gene Aliases
-
Chromosome Location
Chr.1: 273854189 - 273961982 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164103.1 NP_001157575.1 10 - 11 1619 140
NM_031552.2 NP_113740.2 10 - 11 1619 140
XM_006231599.2 XP_006231661.1 10 - 11 1472 140
XM_006231600.2 XP_006231662.1 11 - 12 1719 140
XM_006231601.2 XP_006231663.1 10 - 11 2318 140
XM_006231602.2 XP_006231664.1 10 - 11 2320 140
XM_008760510.1 XP_008758732.1 10 - 11 1462 140
XM_008760511.1 XP_008758733.1 10 - 11 2317 140
GenBank mRNA CO402732.1 - 1 - 2 105 140
U35775.1 - - 1536 140

View Full Product Details