Assay Design



Gene Transcripts

Gene Symbol
Stx5
Entrez Gene ID
65134
Gene Name
syntaxin 5
Gene Aliases
Stx5a
Chromosome Location
Chr.1: 224939497 - 224955658 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031704.2 NP_113892.2 2 - 3 366 69
XM_006230997.2 XP_006231059.1 1 - 2 909 69
GenBank mRNA BC085724.1 - 1 - 2 613 69
BC127489.1 - 2 - 3 366 69
L20822.1 - 1 - 2 196 69
U87971.1 - 2 - 3 366 69

View Full Product Details