Assay Design



Gene Transcripts

Gene Symbol
Psmc1
Entrez Gene ID
117263
Gene Name
proteasome (prosome, macropain) 26S subunit, ATPase, 1
Gene Aliases
s4
Chromosome Location
Chr.6: 124123283 - 124135644 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057123.1 NP_476464.1 5 - 6 608 117
GenBank mRNA BC063157.1 - 6 - 7 620 117
D50696.1 - 5 - 6 608 117
FQ209916.1 - 6 - 7 613 117

View Full Product Details