Assay Design



Gene Transcripts

Gene Symbol
Bnip1
Entrez Gene ID
140932
Gene Name
BCL2/adenovirus E1B interacting protein 1
Gene Aliases
Bnip1l1
Chromosome Location
Chr.10: 16677077 - 16689321 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080897.1 NP_543173.1 5 - 6 503 67
GenBank mRNA AF454391.1 - 5 - 6 503 67
FQ218385.1 - 5 - 6 507 67

View Full Product Details