Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130402.2 | NP_569086.1 | 1 - 2 | 162 | 97 |
XM_008763309.1 | XP_008761531.1 | 2 - 3 | 659 | 97 | |
GenBank mRNA | AF321552.1 | - | 3 - 4 | 348 | 97 |
EF100688.1 | - | 1 - 2 | 162 | 97 |