Assay Design



Gene Transcripts

Gene Symbol
Palm
Entrez Gene ID
170673
Gene Name
paralemmin
Gene Aliases
-
Chromosome Location
Chr.7: 12708024 - 12721055 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130829.1 NP_570842.1 - 685 76
GenBank mRNA AB058889.1 - - 685 76
BC072525.1 - - 726 76

View Full Product Details