Assay Design
Gene Transcripts
- Gene Symbol
- Sval2
- Entrez Gene ID
-
84605
- Gene Name
- seminal vesicle antigen-like 2
- Gene Aliases
-
SLP-M
- Chromosome Location
-
Chr.4: 71340674 - 71344638 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_133291.1 |
NP_579825.1 |
3 - 4 |
310 |
93 |
GenBank mRNA |
AB051831.1 |
- |
3 - 4 |
310 |
93 |
View Full Product Details