Assay Design
Gene Transcripts
- Gene Symbol
- Nmbr
- Entrez Gene ID
-
25264
- Gene Name
- neuromedin B receptor
- Gene Aliases
-
NMB-R
- Chromosome Location
-
Chr.1: 8933770 - 8945240 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012799.1 |
NP_036931.1 |
2 - 3 |
908 |
91 |
GenBank mRNA |
U37058.1 |
- |
2 - 3 |
908 |
91 |
View Full Product Details