Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013200.1 | NP_037332.1 | 14 - 15 | 1779 | 74 |
XM_006242180.2 | XP_006242242.1 | 14 - 15 | 2235 | 74 | |
GenBank mRNA | BC085761.1 | - | 14 - 15 | 1791 | 74 |
D43623.1 | - | 13 - 14 | 1766 | 74 |