Assay Design



Gene Transcripts

Gene Symbol
Kcnk10
Entrez Gene ID
65272
Gene Name
potassium channel, two pore domain subfamily K, member 10
Gene Aliases
TREK2
Chromosome Location
Chr.6: 122417079 - 122547485 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023096.2 NP_075584.1 3 - 4 872 99
XM_008764847.1 XP_008763069.1 3 - 4 1592 99
GenBank mRNA AF196965.1 - 3 - 4 683 99
AF385401.1 - 3 - 4 872 99

View Full Product Details