Assay Design



Gene Transcripts

Gene Symbol
Adh7
Entrez Gene ID
171178
Gene Name
alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide
Gene Aliases
-
Chromosome Location
Chr.2: 243500558 - 243516865 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134329.1 NP_599156.1 5 - 6 620 97
XM_008761493.1 XP_008759715.1 6 - 7 1007 97
GenBank mRNA X98746.1 - 5 - 6 620 97

View Full Product Details