Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138877.1 | NP_620232.1 | 8 - 9 | 773 | 74 |
XM_006242065.2 | XP_006242127.1 | 8 - 9 | 801 | 74 | |
GenBank mRNA | BC062066.1 | - | 8 - 9 | 789 | 74 |
D00636.1 | - | 8 - 9 | 773 | 74 | |
J03867.1 | - | 8 - 9 | 768 | 74 |