Assay Design



Gene Transcripts

Gene Symbol
Pla1a
Entrez Gene ID
85311
Gene Name
phospholipase A1 member A
Gene Aliases
Ps-pla1, Pspla1
Chromosome Location
Chr.11: 64882025 - 64919714 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138882.1 NP_620237.1 8 - 9 1022 116
XM_006248377.2 XP_006248439.1 8 - 9 1431 116
GenBank mRNA BC078727.1 - 8 - 9 1022 116
D88666.1 - 8 - 9 1022 116

View Full Product Details