Assay Design
Gene Transcripts
- Gene Symbol
- Klrh1
- Entrez Gene ID
-
246043
- Gene Name
- killer cell lectin-like receptor subfamily H, member 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 163597634 - 163611383 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139187.2 |
NP_631926.1 |
2 - 3 |
153 |
65 |
GenBank mRNA |
AF416564.2 |
- |
2 - 3 |
153 |
65 |
View Full Product Details