Assay Design



Gene Transcripts

Gene Symbol
Macrod1
Entrez Gene ID
246233
Gene Name
MACRO domain containing 1
Gene Aliases
Lrp16
Chromosome Location
Chr.1: 222310919 - 222451485 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139337.1 NP_647553.1 1 - 2 103 89
XM_006230677.2 XP_006230739.1 1 - 2 437 89
GenBank mRNA AF404762.1 - 1 - 2 103 89
AF419856.1 - 1 - 2 136 89
FQ216840.1 - 1 - 2 270 89

View Full Product Details