Assay Design



Gene Transcripts

Gene Symbol
Gpsm1
Entrez Gene ID
246254
Gene Name
G-protein signaling modulator 1
Gene Aliases
Ags3
Chromosome Location
Chr.3: 3767394 - 3794360 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145469.1 NP_001138941.1 2 - 3 282 78
NM_144745.2 NP_653346.2 12 - 13 1722 78
XM_006233670.2 XP_006233732.1 12 - 13 1826 78
XM_008761620.1 XP_008759842.1 4 - 5 3582 78
GenBank mRNA AF107723.1 - 11 - 12 1846 78
AY136742.1 - 1 - 2 83 78
BC086535.1 - 2 - 3 282 78

View Full Product Details