Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144748.1 | NP_653349.1 | 12 - 13 | 1614 | 77 |
XM_008759677.1 | XP_008757899.1 | 12 - 13 | 1599 | 77 | |
XM_008759678.1 | XP_008757900.1 | 11 - 12 | 1555 | 77 | |
XM_008759679.1 | XP_008757901.1 | 9 - 10 | 1352 | 77 | |
GenBank mRNA | AF062389.1 | - | 12 - 13 | 1614 | 77 |
BC078721.1 | - | 12 - 13 | 1591 | 77 |