Assay Design



Gene Transcripts

Gene Symbol
Dlgap3
Entrez Gene ID
286923
Gene Name
discs, large (Drosophila) homolog-associated protein 3
Gene Aliases
DAP-3, Dap3, SAPAP3
Chromosome Location
Chr.5: 145237589 - 145304536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276304.1 NP_001263233.1 5 - 6 1728 111
XM_008763989.1 XP_008762211.1 6 - 7 3030 111
GenBank mRNA AY530298.1 - 4 - 5 1591 111
AY530299.1 - 4 - 5 1591 111
U67139.1 - 5 - 6 1695 111

View Full Product Details