Assay Design



Gene Transcripts

Gene Symbol
Acaa1a
Entrez Gene ID
24157
Gene Name
acetyl-CoA acyltransferase 1
Gene Aliases
Acaa, Acaa1, Pktaa
Chromosome Location
Chr.8: 128027880 - 128036471 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012489.2 NP_036621.1 2 - 3 369 62
GenBank mRNA BC078905.1 - 2 - 3 293 62
BC089821.1 - 2 - 3 356 62
M32801.1 - 2 - 3 369 62

View Full Product Details