Assay Design



Gene Transcripts

Gene Symbol
Rbp1
Entrez Gene ID
25056
Gene Name
retinol binding protein 1, cellular
Gene Aliases
-
Chromosome Location
Chr.8: 106449321 - 106470842 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012733.4 NP_036865.1 1 - 2 168 77
GenBank mRNA FQ210835.1 - 1 - 2 183 77
M16459.1 - 1 - 2 168 77
M19257.1 - 1 - 2 98 77

View Full Product Details