Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012867.1 | NP_036999.1 | 2 - 3 | 304 | 93 |
GenBank mRNA | FQ215609.1 | - | 2 - 3 | 336 | 93 |
FQ219073.1 | - | 2 - 3 | 336 | 93 | |
FQ220812.1 | - | 2 - 3 | 336 | 93 | |
FQ225610.1 | - | 2 - 3 | 308 | 93 | |
FQ230574.1 | - | 2 - 3 | 308 | 93 | |
FQ232057.1 | - | 2 - 3 | 336 | 93 | |
U72660.1 | - | 2 - 3 | 326 | 93 |