Assay Design



Gene Transcripts

Gene Symbol
Sqle
Entrez Gene ID
29230
Gene Name
squalene epoxidase
Gene Aliases
-
Chromosome Location
Chr.7: 99609929 - 99624803 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017136.2 NP_058832.1 4 - 5 887 72
GenBank mRNA BC097330.1 - 4 - 5 887 72
D37920.1 - 4 - 5 1018 72

View Full Product Details