Assay Design



Gene Transcripts

Gene Symbol
Uhmk1
Entrez Gene ID
246332
Gene Name
U2AF homology motif (UHM) kinase 1
Gene Aliases
KIS, Kist
Chromosome Location
Chr.13: 88521625 - 88536728 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017293.1 NP_058989.1 1 - 2 428 81
GenBank mRNA BC098669.1 - 1 - 2 423 81
U70372.2 - 1 - 2 428 81
X98374.1 - 1 - 2 305 81

View Full Product Details