Assay Design



Gene Transcripts

Gene Symbol
Sept3
Entrez Gene ID
56003
Gene Name
septin 3
Gene Aliases
Sep3
Chromosome Location
Chr.7: 123506535 - 123526542 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019375.1 NP_062248.1 8 - 9 896 73
GenBank mRNA AF111179.1 - 8 - 9 896 73
AF111180.1 - 6 - 7 1303 73

View Full Product Details