Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133286.1 | NP_579820.1 | 4 - 5 | 319 | 72 |
XM_006231436.2 | XP_006231498.1 | 4 - 5 | 511 | 72 | |
XM_006231437.2 | XP_006231499.1 | 5 - 6 | 439 | 72 | |
XM_008760398.1 | XP_008758620.1 | 5 - 6 | 406 | 72 | |
GenBank mRNA | AB079113.1 | - | 4 - 5 | 319 | 72 |