Assay Design
Gene Transcripts
- Gene Symbol
- Serpine1
- Entrez Gene ID
-
24617
- Gene Name
- serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1
- Gene Aliases
-
PAI1A, Pai1, Pai1aa, Planh, RATPAI1A
- Chromosome Location
-
Chr.12: 22641104 - 22651482 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_012620.1 |
NP_036752.1 |
3 - 4 |
615 |
76 |
GenBank mRNA |
M24067.1 |
- |
3 - 4 |
615 |
76 |
View Full Product Details