Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022543.2 | NP_071988.2 | 3 - 4 | 2414 | 86 |
GenBank mRNA | AF223677.1 | - | 3 - 4 | 2521 | 86 |
AY548105.1 | - | 3 - 4 | 2414 | 86 | |
BC070926.1 | - | 2 - 3 | 1752 | 86 |