Assay Design



Gene Transcripts

Gene Symbol
Trpc4
Entrez Gene ID
84494
Gene Name
transient receptor potential cation channel, subfamily C, member 4
Gene Aliases
Trp4, Trrp4
Chromosome Location
Chr.2: 143433183 - 143605757 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083115.1 NP_001076584.1 8 - 9 2124 71
NM_080396.1 NP_536321.1 8 - 9 2124 71
XM_006232356.2 XP_006232418.1 9 - 10 2448 71
GenBank mRNA AB008889.2 - 9 - 10 2170 71
AF288407.1 - 8 - 9 2140 71
AF288408.1 - 8 - 9 2140 71
AF421363.1 - 8 - 9 2124 71
AF421364.1 - 8 - 9 2124 71
AF421365.1 - 8 - 9 2124 71
AF421366.1 - 8 - 9 2056 71
AF421367.1 - 8 - 9 2071 71
AF421368.1 - 8 - 9 2113 71
FQ228645.1 - 9 - 10 2343 71

View Full Product Details