Assay Design



Gene Transcripts

Gene Symbol
Bckdk
Entrez Gene ID
29603
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
-
Chromosome Location
Chr.1: 199351628 - 199356299 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019244.2 NP_062117.2 10 - 11 1355 77
GenBank mRNA BM392152.1 - 1 - 2 77
M93271.1 - - 1220 77
U27456.1 - 9 - 10 1009 77

View Full Product Details