Assay Design



Gene Transcripts

Gene Symbol
Chp2
Entrez Gene ID
308965
Gene Name
calcineurin-like EF hand protein 2
Gene Aliases
chp-2
Chromosome Location
Chr.1: 192159071 - 192173228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182738.1 NP_877402.1 2 - 3 182 77
XM_008759745.1 XP_008757967.1 3 - 4 360 77
XM_008759746.1 XP_008757968.1 3 - 4 330 77
GenBank mRNA AB086189.1 - 2 - 3 182 77

View Full Product Details