Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031556.3 | NP_113744.2 | 2 - 3 | 347 | 64 |
NM_133651.2 | NP_598412.2 | 1 - 2 | 339 | 64 | |
XM_006236130.2 | XP_006236192.1 | 2 - 3 | 319 | 64 | |
XM_006236131.1 | XP_006236193.1 | 2 - 3 | 286 | 64 | |
GenBank mRNA | AF439778.1 | - | 2 - 3 | 192 | 64 |
AF439779.1 | - | 1 - 2 | 99 | 64 | |
BC078744.1 | - | 2 - 3 | 226 | 64 | |
BC104689.1 | - | 1 - 2 | 162 | 64 | |
BC128717.1 | - | 1 - 2 | 169 | 64 | |
BC161826.1 | - | 1 - 2 | 332 | 64 | |
CB698936.1 | - | 2 - 3 | 241 | 64 | |
CK364516.1 | - | 1 - 2 | 339 | 64 | |
FM039194.1 | - | 2 - 3 | 347 | 64 | |
Z46614.1 | - | 2 - 3 | 220 | 64 |