Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017150.1 | NP_058846.1 | 3 - 4 | 142 | 66 |
GenBank mRNA | BC086404.1 | - | 3 - 4 | 136 | 66 |
FQ221647.1 | - | 3 - 4 | 149 | 66 | |
FQ222035.1 | - | 3 - 4 | 164 | 66 | |
FQ229056.1 | - | 3 - 4 | 147 | 66 | |
FQ229057.1 | - | 3 - 4 | 147 | 66 | |
X60744.1 | - | 3 - 4 | 171 | 66 | |
X68283.1 | - | 3 - 4 | 142 | 66 |