Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017271.2 | NP_058967.1 | 7 - 8 | 1034 | 99 |
GenBank mRNA | BC065581.1 | - | 7 - 8 | 1020 | 99 |
X82445.1 | - | 7 - 8 | 1034 | 99 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_008773882.1 | XP_008772104.1 | 6 - 7 | 839 | 99 |