Assay Design
Gene Transcripts
- Gene Symbol
- Hes5
- Entrez Gene ID
-
79225
- Gene Name
- hes family bHLH transcription factor 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.5: 172364421 - 172365188 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_024383.1 |
NP_077359.1 |
2 - 3 |
274 |
103 |
GenBank mRNA |
D12516.1 |
- |
2 - 3 |
274 |
103 |
View Full Product Details