Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053345.1 | NP_445797.1 | 3 - 3 | 288 | 91 |
XM_006243380.2 | XP_006243442.1 | 4 - 4 | 438 | 91 | |
GenBank mRNA | AF000944.1 | - | 3 - 3 | 288 | 91 |
BC168727.1 | - | 5 - 5 | 472 | 91 | |
FQ209546.1 | - | - | 443 | 91 | |
FQ212213.1 | - | 4 - 4 | 423 | 91 | |
FQ221947.1 | - | 4 - 4 | 429 | 91 |