Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057119.1 | NP_476460.1 | 1 - 2 | 176 | 69 |
XM_006248536.2 | XP_006248598.1 | 1 - 2 | 217 | 69 | |
GenBank mRNA | BC070948.1 | - | 1 - 2 | 176 | 69 |
D49708.1 | - | 1 - 2 | 176 | 69 | |
FQ212414.1 | - | 1 - 2 | 190 | 69 |