Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139253.1 | NP_640346.1 | 2 - 3 | 223 | 62 |
XM_006239710.2 | XP_006239772.1 | 2 - 3 | 262 | 62 | |
XM_006239711.2 | XP_006239773.1 | 2 - 3 | 283 | 62 | |
GenBank mRNA | AF475282.1 | - | 2 - 3 | 223 | 62 |
BC071179.1 | - | 2 - 3 | 217 | 62 |