Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013139.1 | NP_037271.1 | 1 - 2 | 141 | 100 |
XM_008772711.1 | XP_008770933.1 | 1 - 2 | 152 | 100 | |
GenBank mRNA | M33333.1 | - | 1 - 2 | 100 | 100 |
M58370.1 | - | 1 - 2 | 141 | 100 |