Assay Design



Gene Transcripts

Gene Symbol
Smek1
Entrez Gene ID
314388
Gene Name
SMEK homolog 1, suppressor of mek1 (Dictyostelium)
Gene Aliases
RGD1309059
Chromosome Location
Chr.6: 125040803 - 125083496 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108050.2 NP_001101520.2 8 - 9 1821 81
NM_001271174.1 NP_001258103.1 8 - 9 2118 81
GenBank mRNA EV776435.1 - 3 - 4 307 81

View Full Product Details