Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127490.1 | NP_001120962.1 | 6 - 7 | 1189 | 65 |
XM_006243326.2 | XP_006243388.1 | 7 - 8 | 1102 | 65 | |
XM_008766291.1 | XP_008764513.1 | 8 - 9 | 1066 | 65 | |
XM_008766292.1 | XP_008764514.1 | 8 - 9 | 750 | 65 | |
GenBank mRNA | BC158676.1 | - | 6 - 7 | 620 | 65 |