Assay Design



Gene Transcripts

Gene Symbol
Vgll3
Entrez Gene ID
498038
Gene Name
vestigial-like family member 3
Gene Aliases
RGD1560030
Chromosome Location
Chr.11: 3119505 - 3168888 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247981.2 XP_006248043.1 1 - 2 543 79

View Full Product Details