Assay Design



Gene Transcripts

Gene Symbol
Rora
Entrez Gene ID
300807
Gene Name
RAR-related orphan receptor A
Gene Aliases
-
Chromosome Location
Chr.8: 75515882 - 75616477 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766408.1 XP_008764630.1 2 - 3 562 71
XM_008766409.1 XP_008764631.1 2 - 3 563 71
XM_008766410.1 XP_008764632.1 2 - 3 218 71

View Full Product Details