Assay Design



Gene Transcripts

Gene Symbol
Rpusd4
Entrez Gene ID
315550
Gene Name
RNA pseudouridylate synthase domain containing 4
Gene Aliases
RGD1311268
Chromosome Location
Chr.8: 36467706 - 36477190 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025284.1 NP_001020455.1 3 - 4 563 60
GenBank mRNA BC098023.1 - 3 - 4 563 60

View Full Product Details