Assay Design



Gene Transcripts

Gene Symbol
Smap2
Entrez Gene ID
298500
Gene Name
small ArfGAP2
Gene Aliases
RGD1308418, Smap1l
Chromosome Location
Chr.5: 139885321 - 139931366 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100669.1 NP_001094139.1 2 - 3 607 63
GenBank mRNA BC161927.1 - 2 - 3 601 63

View Full Product Details