Assay Design
Gene Transcripts
- Gene Symbol
- Kat6a
- Entrez Gene ID
-
306571
- Gene Name
- K(lysine) acetyltransferase 6A
- Gene Aliases
-
Moz, Myst3, Runxbp2, Znf220
- Chromosome Location
-
Chr.16: 73942669 - 74022971 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB195309.1 |
- |
11 - 12 |
1970 |
63 |
View Full Product Details